siRNA Details
| Gene Symbol: Coro1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301374.1 | NP_001288303.1 | 8 | 1184 | ||
| NM_009898.3 | NP_034028.1 | 8 | 1097 | |||
| GenBank mRNA | AF047388.1 | - | 7 | 931 | ||
| AF143955.1 | - | 8 | 1068 | |||
| AF495468.1 | - | 8 | 1035 | |||
| AK040885.1 | - | 8 | 1041 | |||
| AK075799.1 | - | 8 | 1033 | |||
| AK146290.1 | - | 8 | 1035 | |||
| AK150962.1 | - | 8 | 1039 | |||
| AK151755.1 | - | 8 | 1039 | |||
| AK155527.1 | - | 8 | 1040 | |||
| AK155860.1 | - | 8 | 1035 | |||
| AK165296.1 | - | 8 | 1039 | |||
| AK172447.1 | - | 8 | 1035 | |||
| BC002136.1 | - | 8 | 1025 | 3484185 | ||
| BC053398.1 | - | 8 | 1015 | 30076939 | ||

Coro1a
Genomic Map