siRNA Details
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF033566.1 | - | 7 | 809 | ||
AK013974.2 | - | 5 | 564 | |||
AK161739.1 | - | 3 | 1818 | |||
AY653308.1 | - | 6 | 699 | |||
BC002220.1 | - | 3 | 365 | 3490616 | ||
BC012675.1 | - | 7 | 831 | 3982920 | ||
CT010293.1 | - | 6 | 731 |
