siRNA Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009906.5 | NP_034036.1 | 7 | 724 | ||
| GenBank mRNA | AF111172.1 | - | 7 | 732 | ||
| AJ011912.1 | - | 7 | 718 | |||
| AK002418.1 | - | 7 | 743 | |||
| AK048279.1 | - | 7 | 725 | |||
| AK083832.1 | - | 6 | 1989 | |||
| AK146759.1 | - | 7 | 717 | |||
| AK155021.1 | - | 7 | 743 | |||
| AK165669.1 | - | 7 | 749 | |||
| AK166648.1 | - | 5 | 518 | |||
| AK170781.1 | - | 7 | 720 | |||
| BC024820.1 | - | 7 | 726 | 5360558 | ||

Tpp1
Genomic Map