siRNA Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009906.5 | NP_034036.1 | 3, 4 | 251 | ||
| GenBank mRNA | AF111172.1 | - | 3, 4 | 259 | ||
| AJ011912.1 | - | 3, 4 | 245 | |||
| AK002418.1 | - | 3, 4 | 270 | |||
| AK048279.1 | - | 3, 4 | 252 | |||
| AK146759.1 | - | 3, 4 | 244 | |||
| AK155021.1 | - | 3, 4 | 270 | |||
| AK165669.1 | - | 3, 4 | 276 | |||
| AK166648.1 | - | 1, 2 | 45 | |||
| AK169924.1 | - | 3, 4 | 258 | |||
| AK170781.1 | - | 3, 4 | 247 | |||
| BC024820.1 | - | 3, 4 | 253 | 5360558 | ||

Tpp1
Genomic Map