siRNA Details
| Gene Symbol: Clock | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289826.1 | NP_001276755.1 | 11 | 1125 | ||
| NM_007715.6 | NP_031741.1 | 11 | 1168 | |||
| GenBank mRNA | AB221635.1 | - | 8 | 780 | ||
| AF000998.1 | - | 11 | 1168 | |||
| AK030352.1 | - | 11 | 1068 | |||
| AK040337.1 | - | 11 | 1106 | |||
| AK129118.1 | - | 11 | 1125 | |||
| BC129886.1 | - | 9 | 877 | |||

Clock
Genomic Map