siRNA Details
| Gene Symbol: Ccr1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007718.3 | NP_031744.3 | 1 | 281 | ||
| GenBank mRNA | BC127143.1 | - | 1 | 281 | 40051964 | |
| BC128466.1 | - | 1 | 281 | 40051965 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr1l1| Gene Symbol: Ccr1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007718.3 | NP_031744.3 | 1 | 281 | ||
| GenBank mRNA | BC127143.1 | - | 1 | 281 | 40051964 | |
| BC128466.1 | - | 1 | 281 | 40051965 | ||