siRNA Details
| Gene Symbol: Abcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013806.2 | NP_038834.2 | 23 | 3331 | ||
| GenBank mRNA | AF227274.1 | - | 23 | 3235 | ||
| AF282772.1 | - | 23 | 3235 | |||
| AF282773.1 | - | 23 | 3235 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcc2| Gene Symbol: Abcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013806.2 | NP_038834.2 | 23 | 3331 | ||
| GenBank mRNA | AF227274.1 | - | 23 | 3235 | ||
| AF282772.1 | - | 23 | 3235 | |||
| AF282773.1 | - | 23 | 3235 | |||