siRNA Details
| Gene Symbol: Cnga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282010.1 | NP_001268939.1 | 8 | 1305 | ||
| NM_009918.2 | NP_034048.1 | 6 | 1250 | |||
| GenBank mRNA | AJ243933.1 | - | 6 | 1259 | ||
| BC035272.1 | - | 6 | 1194 | 5357087 | ||
| BC049145.1 | - | 6 | 1193 | 5357087 | ||

Cnga3
Genomic Map