siRNA Details
| Gene Symbol: Cnih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009919.2 | NP_034049.2 | 4 | 468 | ||
| GenBank mRNA | AF022811.1 | - | 4 | 459 | ||
| AK002293.1 | - | 4 | 436 | |||
| AK011200.1 | - | 4 | 421 | |||
| AK152432.1 | - | 4 | 419 | |||
| AK154479.1 | - | 4 | 419 | |||
| AK168154.1 | - | 4 | 419 | |||
| BC034868.1 | - | 4 | 428 | 2938052 | ||
| DV653213.1 | - | 4 | 522 | |||

Cnih1
Genomic Map