siRNA Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013807.2 | NP_038835.2 | 3 | 493 | ||
| GenBank mRNA | AK156754.1 | - | 3 | 505 | ||
| AK209626.1 | - | 1 | 45 | |||
| BC031180.1 | - | 3 | 485 | 4988360 | ||
| BC063051.1 | - | 3 | 493 | 5718130 | ||
| U21392.1 | - | 3 | 340 | |||

Plk3
Genomic Map