siRNA Details
| Gene Symbol: Camp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009921.2 | NP_034051.2 | 2 | 294 | ||
| GenBank mRNA | AK158005.1 | - | 2 | 286 | ||
| BC120752.1 | - | 2 | 291 | 40129675 | ||
| BC120754.1 | - | 2 | 294 | 40129677 | ||
| BY582734.1 | - | 1 | 95 | |||
| U43409.1 | - | 2 | 287 | |||
| X94353.1 | - | 2 | 286 | |||

Camp
Genomic Map