siRNA Details
| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009922.4 | NP_034052.3 | 2, 3 | 281 | ||
| GenBank mRNA | AK035548.1 | - | 2, 3 | 254 | ||
| BY326336.1 | - | 2 | 319 | |||
| Z19542.1 | - | 2, 3 | 275 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnn1| Gene Symbol: Cnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009922.4 | NP_034052.3 | 2, 3 | 281 | ||
| GenBank mRNA | AK035548.1 | - | 2, 3 | 254 | ||
| BY326336.1 | - | 2 | 319 | |||
| Z19542.1 | - | 2, 3 | 275 | |||