siRNA Details
| Gene Symbol: Cnr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036658.1 | - | 2 | 779 | ||
| AK037898.1 | - | 2 | 889 | |||
| AK134109.1 | - | 2 | 815 | |||
| BC024052.1 | - | 2 | 744 | 5098695 | ||
| X86405.1 | - | 2 | 732 | |||
| X93168.1 | - | 2 | 800 | |||

Cnr2
Genomic Map