siRNA Details
| Gene Symbol: Cntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159647.1 | NP_001153119.1 | 11 | 1556 | ||
| NM_001159648.1 | NP_001153120.1 | 11 | 1410 | |||
| NM_007727.2 | NP_031753.1 | 11 | 1459 | |||
| GenBank mRNA | BC066864.1 | - | 11 | 1482 | 30099512 | |
| X14943.1 | - | 11 | 1429 | |||
More Information
Other Species Targeted
Set Membership: |
|

Cntn1
Genomic Map