siRNA Details
| Gene Symbol: Cntn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159647.1 | NP_001153119.1 | 22, 23 | 3213 | ||
| NM_001159648.1 | NP_001153120.1 | 22, 23 | 3067 | |||
| NM_007727.2 | NP_031753.1 | 22, 23 | 3116 | |||
| GenBank mRNA | AK045710.1 | - | 3, 4 | 406 | ||
| AK076273.1 | - | 8, 9 | 1120 | |||
| BC066864.1 | - | 22, 23 | 3139 | 30099512 | ||
| X14943.1 | - | 22, 23 | 3086 | |||

Cntn1
Genomic Map