siRNA Details
| Gene Symbol: Hps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146323.1 | NP_001139795.1 | 10 | 1368 | ||
| NM_080634.4 | NP_542365.3 | 10 | 1764 | |||
| GenBank mRNA | AF393747.1 | - | 10 | 1753 | ||
| AF393780.1 | - | 10 | 1756 | |||
| AK034377.1 | - | 10 | 1735 | |||
| AK036663.1 | - | 10 | 1772 | |||
| AK037829.1 | - | 9 | 1108 | |||
| AK155497.1 | - | 10 | 1731 | |||
| AK166194.1 | - | 10 | 1749 | |||
| AK169855.1 | - | 9 | 1836 | |||
| AK170581.1 | - | 10 | 1280 | |||
| AK171997.1 | - | 10 | 1356 | |||
| BC058235.1 | - | 10 | 1738 | 6307400 | ||

Hps3
Genomic Map