siRNA Details
| Gene Symbol: HIST1H2BD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021063.3 | NP_066407.1 | 1 | 121 | ||
| NM_138720.2 | NP_619790.1 | 1 | 121 | |||
| XM_005249039.3 | XP_005249096.1 | 1 | 257 | |||
| GenBank mRNA | AJ223353.1 | - | 1 | 100 | ||
| AK292712.1 | - | 1 | 117 | |||
| BC002842.2 | - | 1 | 97 | 3659807 | ||
| BC096122.3 | - | 1 | 72 | 40002354 | ||
| BE245851.1 | - | 1 | 121 | |||
| BM153714.1 | - | 1 | 121 | |||

HIST1H2BD
Genomic Map