siRNA Details
| Gene Symbol: Col4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009932.3 | NP_034062.3 | 48 | 5321 | ||
| GenBank mRNA | AK075619.1 | - | 1 | 47 | ||
| AY375463.1 | - | 3 | 550 | |||
| AY502947.1 | - | 2 | 265 | |||
| BC013560.1 | - | 6 | 978 | |||
| BC138040.1 | - | 47 | 5033 | 8861059 | ||
| J04695.1 | - | 48 | 5126 | |||
| M15833.1 | - | 3 | 550 | |||
| X04647.1 | - | 16 | 2083 | |||

Col4a2
Genomic Map