siRNA Details
| Gene Symbol: Col4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009932.3 | NP_034062.3 | 42 | 4236 | ||
| GenBank mRNA | BC138040.1 | - | 41 | 3948 | 8861059 | |
| J04695.1 | - | 42 | 4041 | |||
| X04410.1 | - | 9 | 785 | |||
| X04647.1 | - | 10 | 998 | |||

Col4a2
Genomic Map