siRNA Details
| Gene Symbol: Col4a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163155.1 | NP_001156627.1 | 13 | 1050 | ||
| NM_007736.4 | NP_031762.2 | 14 | 878 | |||
| GenBank mRNA | AB041350.1 | - | 13 | 908 | ||
| AK133179.1 | - | 14 | 879 | |||
| BC082788.1 | - | 13 | 889 | 5343488 | ||

Col4a5
Genomic Map