siRNA Details
| Gene Symbol: Col8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007739.2 | NP_031765.2 | 3 | 262 | ||
| GenBank mRNA | AK018742.1 | - | 3 | 241 | ||
| AK029203.1 | - | 3 | 237 | |||
| AK032048.1 | - | 3 | 241 | |||
| AK046370.1 | - | 3 | 241 | |||
| AK159542.1 | - | 3 | 237 | |||
| BC011061.1 | - | 3 | 229 | 3674027 | ||
| BC058281.1 | - | 3 | 245 | |||
| BY289783.1 | - | 3 | 263 | |||

Col8a1
Genomic Map