siRNA Details
| Gene Symbol: Col1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007742.3 | NP_031768.2 | 51 | 4342 | ||
| GenBank mRNA | AK132180.1 | - | 51 | 4365 | ||
| AK159283.1 | - | 51 | 4359 | |||
| AK159285.1 | - | 51 | 4365 | |||
| BC003198.1 | - | 14 | 1652 | |||
| BC042642.1 | - | 44 | 3912 | |||
| BC050014.1 | - | 51 | 4342 | 4225038 | ||
| BC059281.1 | - | 44 | 3668 | 3992363 | ||
| U03419.1 | - | 5 | 859 | |||
| U08020.1 | - | 51 | 4243 | |||

Col1a1
Genomic Map