siRNA Details
| Gene Symbol: Copa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009938.4 | NP_034068.3 | 20, 21 | 2549 | ||
| GenBank mRNA | AK043479.1 | - | 20, 21 | 2275 | ||
| AK052724.1 | - | 2, 3 | 271 | |||
| BC024070.1 | - | 20, 21 | 2202 | 5097228 | ||
| BC025896.1 | - | 2, 3 | 236 | |||
| BC047429.1 | - | 20, 21 | 2240 | 3498656 | ||
| BC082785.1 | - | 16, 17 | 1889 | |||

Copa
Genomic Map