siRNA Details
| Gene Symbol: Copa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009938.4 | NP_034068.3 | 26 | 3126 | ||
| GenBank mRNA | AK032071.1 | - | 4 | 1079 | ||
| AK043479.1 | - | 26 | 2852 | |||
| BC024070.1 | - | 26 | 2779 | 5097228 | ||
| BC025896.1 | - | 8 | 813 | |||
| BC047429.1 | - | 26 | 2817 | 3498656 | ||
| BC082785.1 | - | 22 | 2466 | |||

Copa
Genomic Map