siRNA Details
| Gene Symbol: Cox7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009945.3 | NP_034075.2 | 4 | 608 | ||
| GenBank mRNA | AK007875.1 | - | 4 | 485 | ||
| AK019210.1 | - | 4 | 463 | |||
| AK153173.1 | - | 4 | 485 | |||
| CN670202.1 | - | 4 | 471 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cox7a2| Gene Symbol: Cox7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009945.3 | NP_034075.2 | 4 | 608 | ||
| GenBank mRNA | AK007875.1 | - | 4 | 485 | ||
| AK019210.1 | - | 4 | 463 | |||
| AK153173.1 | - | 4 | 485 | |||
| CN670202.1 | - | 4 | 471 | |||