siRNA Details
| Gene Symbol: Cox7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007749.3 | NP_031775.1 | 3 | 357 | ||
| GenBank mRNA | AK002494.1 | - | 3 | 324 | ||
| AK027989.1 | - | 3 | 319 | |||
| AK131871.1 | - | 3 | 353 | |||
| AK153444.1 | - | 3 | 329 | |||
| AV056265.1 | - | 3 | 197 | |||
| BB696274.1 | - | 3 | 368 | |||
| BC086792.1 | - | 3 | 329 | 6411273 | ||

Cox7c
Genomic Map