siRNA Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276248.1 | NP_001263177.1 | 17 | 3132 | ||
| NM_001276250.1 | NP_001263179.1 | 17 | 3132 | |||
| NM_007752.3 | NP_031778.2 | 17 | 3135 | |||
| GenBank mRNA | AK014835.1 | - | 17 | 3116 | ||
| AK043248.1 | - | 17 | 3077 | |||
| AK080701.1 | - | 9 | 1231 | |||
| AK160035.1 | - | 13 | 2061 | |||
| BC062957.1 | - | 17 | 3135 | 30547760 | ||
| DQ336396.1 | - | 17 | 3133 | |||
| U49430.1 | - | 17 | 3013 | |||

Cp
Genomic Map