siRNA Details
| Gene Symbol: Cpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007753.2 | NP_031779.1 | 11 | 1220 | ||
| GenBank mRNA | AK088906.1 | - | 11 | 1221 | ||
| BC108409.1 | - | 11 | 1210 | |||
| J05118.1 | - | 11 | 1216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpa3| Gene Symbol: Cpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007753.2 | NP_031779.1 | 11 | 1220 | ||
| GenBank mRNA | AK088906.1 | - | 11 | 1221 | ||
| BC108409.1 | - | 11 | 1210 | |||
| J05118.1 | - | 11 | 1216 | |||