siRNA Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007762.4 | NP_031788.1 | 9, 10 | 1038 | ||
| GenBank mRNA | AB375515.1 | - | 9, 10 | 848 | ||
| AF369656.1 | - | 1, 2 | 82 | |||
| AF483484.1 | - | 9, 10 | 848 | |||
| AF483485.1 | - | 9, 10 | 848 | |||
| AK144430.1 | - | 9, 10 | 1106 | |||
| AK158806.1 | - | 9, 10 | 1039 | |||
| BC103675.1 | - | 9, 10 | 866 | 40046992 | ||
| BC104735.1 | - | 10, 11 | 866 | 40046995 | ||
| BC105673.1 | - | 10, 11 | 942 | 40046994 | ||
| BC105675.1 | - | 9, 10 | 866 | 40046993 | ||
| FJ668671.1 | - | 8, 9 | 728 | |||
| X72305.1 | - | 9, 10 | 879 | |||
More Information
Other Species Targeted
Set Membership: |
|

Crhr1
Genomic Map