siRNA Details
| Gene Symbol: Dmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007769.2 | NP_031795.2 | 45 | 5272 | ||
| GenBank mRNA | AB005909.1 | - | 46 | 5301 | ||
| BC022638.1 | - | 4 | 684 | |||
| BC049835.1 | - | 45 | 5272 | 6396511 | ||
| FJ234411.1 | - | 7 | 1204 | |||
| U37438.1 | - | 5682 | ||||

Dmbt1
Genomic Map