siRNA Details
| Gene Symbol: Cryba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009965.2 | NP_034095.1 | 3 | 177 | ||
| GenBank mRNA | AJ239052.1 | - | 3 | 177 | ||
| BC107342.2 | - | 3 | 177 | 40055280 | ||
| V00728.1 | - | 1 | 69 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryba1| Gene Symbol: Cryba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009965.2 | NP_034095.1 | 3 | 177 | ||
| GenBank mRNA | AJ239052.1 | - | 3 | 177 | ||
| BC107342.2 | - | 3 | 177 | 40055280 | ||
| V00728.1 | - | 1 | 69 | |||