siRNA Details
| Gene Symbol: Cryba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021541.3 | NP_067516.1 | 4 | 435 | ||
| GenBank mRNA | AJ272227.1 | - | 4 | 427 | ||
| AK021374.1 | - | 4 | 409 | |||
| AY160973.1 | - | 4 | 400 | |||
| BC052746.1 | - | 4 | 398 | 6824801 | ||
| BC056989.1 | - | 4 | 389 | 6824245 | ||
| CF740624.1 | - | 4 | 434 | |||

Cryba2
Genomic Map