siRNA Details
| Gene Symbol: Crybb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023695.2 | NP_076184.1 | 2, 3 | 219 | ||
| GenBank mRNA | AF106853.1 | - | 2, 3 | 219 | ||
| AK014012.1 | - | 2, 3 | 221 | |||
| AK053869.1 | - | 2, 3 | 220 | |||
| AK076443.1 | - | 2, 3 | 220 | |||
| BC029012.1 | - | 1, 2 | 203 | 5358714 | ||

Crybb1
Genomic Map