siRNA Details
| Gene Symbol: HADH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184705.2 | NP_001171634.2 | 2 | 382 | ||
| NM_005327.4 | NP_005318.3 | 2 | 382 | |||
| XM_005262972.2 | XP_005263029.1 | 2 | 314 | |||
| GenBank mRNA | AF001902.1 | - | 2 | 146 | ||
| AF001903.1 | - | 2 | 382 | |||
| AK096018.1 | - | 2 | 314 | |||
| AK314444.1 | - | 2 | 243 | |||
| BC000306.2 | - | 2 | 190 | 2820507 | ||
| DA825380.1 | - | 2 | 312 | |||
| X96752.1 | - | 2 | 250 | |||

HADH
Genomic Map