siRNA Details
| Gene Symbol: Cryz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009968.3 | NP_034098.1 | 6 | 1176 | ||
| GenBank mRNA | AK098082.1 | - | 6 | 660 | ||
| AK133820.1 | - | 6 | 1177 | |||
| AK134281.1 | - | 6 | 661 | |||
| AK188548.1 | - | 3 | 183 | |||
| BC003800.1 | - | 6 | 668 | 3492859 | ||
| BC043076.1 | - | 6 | 637 | 6404996 | ||
| D78646.1 | - | 3 | 203 | |||
| S70056.1 | - | 6 | 688 | |||

Cryz
Genomic Map