siRNA Details
| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026444.3 | NP_080720.1 | 9 | 1053 | ||
| GenBank mRNA | AB056479.1 | - | 9 | 967 | ||
| AK010298.1 | - | 9 | 1140 | |||
| AK012104.1 | - | 3 | 231 | |||
| AK012151.1 | - | 9 | 1084 | |||
| AK145643.1 | - | 9 | 1085 | |||
| AK149990.1 | - | 9 | 1152 | |||
| AK163273.1 | - | 9 | 1078 | |||
| AK166814.1 | - | 9 | 1088 | |||
| AK167125.1 | - | 9 | 1085 | |||
| BC013554.1 | - | 9 | 1068 | 4237940 | ||
| BC029754.1 | - | 9 | 1053 | 2655044 | ||
| DQ403125.1 | - | 3 | 301 | |||

Cs
Genomic Map