siRNA Details
| Gene Symbol: Cs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026444.3 | NP_080720.1 | 3 | 204 | ||
| GenBank mRNA | AB056479.1 | - | 3 | 118 | ||
| AK010298.1 | - | 3 | 293 | |||
| AK012151.1 | - | 3 | 235 | |||
| AK145643.1 | - | 3 | 236 | |||
| AK149990.1 | - | 3 | 303 | |||
| AK163273.1 | - | 3 | 229 | |||
| AK166814.1 | - | 3 | 239 | |||
| AK167125.1 | - | 3 | 236 | |||
| BC013554.1 | - | 3 | 219 | 4237940 | ||
| BC029754.1 | - | 3 | 204 | 2655044 | ||

Cs
Genomic Map