siRNA Details
| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009970.2 | NP_034100.2 | 12 | 1670 | ||
| GenBank mRNA | AK135114.1 | - | 5 | 1652 | ||
| M85078.1 | - | 12 | 1611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csf2ra| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009970.2 | NP_034100.2 | 12 | 1670 | ||
| GenBank mRNA | AK135114.1 | - | 5 | 1652 | ||
| M85078.1 | - | 12 | 1611 | |||