siRNA Details
| Gene Symbol: Csf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009971.1 | NP_034101.1 | 5 | 1052 | ||
| GenBank mRNA | AK145177.1 | - | 5 | 1017 | ||
| M13926.1 | - | 5 | 1052 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csf3| Gene Symbol: Csf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009971.1 | NP_034101.1 | 5 | 1052 | ||
| GenBank mRNA | AK145177.1 | - | 5 | 1017 | ||
| M13926.1 | - | 5 | 1052 | |||