siRNA Details
Gene Symbol: Csf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009971.1 | NP_034101.1 | 5 | 1192 | ||
GenBank mRNA | AK145177.1 | - | 5 | 1157 | ||
M13926.1 | - | 5 | 1192 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009971.1 | NP_034101.1 | 5 | 1192 | ||
GenBank mRNA | AK145177.1 | - | 5 | 1157 | ||
M13926.1 | - | 5 | 1192 |