siRNA Details
Gene Symbol: Csn1s2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142688.1 | - | 8 | 283 | ||
AK164738.1 | - | 8 | 287 | |||
BC139326.1 | - | 8 | 263 | 8862348 | ||
D10215.1 | - | 8 | 288 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csn1s2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142688.1 | - | 8 | 283 | ||
AK164738.1 | - | 8 | 287 | |||
BC139326.1 | - | 8 | 263 | 8862348 | ||
D10215.1 | - | 8 | 288 |