siRNA Details
| Gene Symbol: Csnk2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009974.3 | NP_034104.1 | 8 | 1012 | ||
| GenBank mRNA | AF012251.1 | - | 8 | 737 | ||
| AJ001420.1 | - | 8 | 982 | |||
| AK002927.1 | - | 8 | 1020 | |||
| AK013008.1 | - | 2 | 52 | |||
| AK030035.1 | - | 2 | 52 | |||
| AK076042.1 | - | 8 | 973 | |||
| BC047143.1 | - | 8 | 1002 | |||
| BC057862.1 | - | 8 | 672 | 5316132 | ||

Csnk2a2
Genomic Map