siRNA Details
| Gene Symbol: Ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007801.2 | NP_031827.2 | 9 | 947 | ||
| GenBank mRNA | AK017638.1 | - | 9 | 679 | ||
| AK149949.1 | - | 9 | 668 | |||
| AK150583.1 | - | 9 | 676 | |||
| AK151877.1 | - | 9 | 688 | |||
| AK157376.1 | - | 9 | 721 | |||
| AK160026.1 | - | 9 | 678 | |||
| AK167498.1 | - | 8 | 572 | |||
| BC006878.1 | - | 9 | 687 | 2647229 | ||
| U06119.1 | - | 9 | 711 | |||
| Y18464.1 | - | 4 | 259 | |||

Ctsh
Genomic Map