siRNA Details
| Gene Symbol: Ctss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267695.2 | NP_001254624.2 | 3 | 376 | ||
| NM_021281.3 | NP_067256.4 | 3 | 373 | |||
| GenBank mRNA | AF038546.1 | - | 3 | 315 | ||
| AJ002386.1 | - | 3 | 328 | |||
| AK028366.1 | - | 3 | 375 | |||
| AK150274.1 | - | 3 | 307 | |||
| AK150758.1 | - | 2 | 219 | |||
| AK150842.1 | - | 2 | 219 | |||
| AK152191.1 | - | 3 | 326 | |||
| AK152618.1 | - | 2 | 219 | |||
| BC002125.1 | - | 3 | 282 | 3495719 | ||
| BC011104.1 | - | 3 | 290 | |||

Ctss
Genomic Map