siRNA Details
| Gene Symbol: Cux1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291233.1 | NP_001278162.1 | 5 | 942 | ||
| NM_001291234.1 | NP_001278163.1 | 5 | 653 | |||
| NM_001291238.1 | NP_001278167.1 | 5 | 653 | |||
| NM_001291239.1 | NP_001278168.1 | 5 | 942 | |||
| NM_001291240.1 | NP_001278169.1 | 5 | 942 | |||
| NM_009986.4 | NP_034116.3 | 5 | 653 | |||
| NM_198602.3 | NP_941004.2 | 5 | 653 | |||
| GenBank mRNA | AB041595.1 | - | 5 | 789 | ||
| AF004225.1 | - | 1 | 12 | |||
| AK098096.1 | - | 5 | 416 | |||
| AK133543.1 | - | 5 | 416 | |||
| AK144932.1 | - | 5 | 1313 | |||
| AK220173.1 | - | 5 | 409 | |||
| AY037807.1 | - | 5 | 383 | |||
| BC014289.1 | - | 5 | 422 | |||
| BC054370.1 | - | 5 | 376 | |||
| BC090847.1 | - | 5 | 425 | 30547458 | ||
| CA317682.1 | - | 5 | 608 | |||
| U68542.1 | - | 6 | 427 | |||

Cux1
Genomic Map