siRNA Details
| Gene Symbol: Cyb561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007805.4 | NP_031831.2 | 4 | 418 | ||
| GenBank mRNA | AK014395.2 | - | 4 | 419 | ||
| AK028780.1 | - | 3 | 292 | |||
| AK158978.1 | - | 5 | 749 | |||
| AK169711.1 | - | 4 | 407 | |||
| BC006732.1 | - | 4 | 458 | 3963612 | ||
| U16297.1 | - | 4 | 438 | |||

Cyb561
Genomic Map