siRNA Details
| Gene Symbol: Cyp11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019779.3 | NP_062753.3 | 5 | 867 | ||
| GenBank mRNA | AF195119.1 | - | 5 | 867 | ||
| AK077945.1 | - | 4 | 1787 | |||
| BC068264.1 | - | 5 | 886 | 30014890 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp11a1| Gene Symbol: Cyp11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019779.3 | NP_062753.3 | 5 | 867 | ||
| GenBank mRNA | AF195119.1 | - | 5 | 867 | ||
| AK077945.1 | - | 4 | 1787 | |||
| BC068264.1 | - | 5 | 886 | 30014890 | ||