siRNA Details
| Gene Symbol: HBD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000519.3 | NP_000510.1 | 3 | 684 | ||
| GenBank mRNA | AY034468.1 | - | 3 | 684 | ||
| BC070282.1 | - | 3 | 540 | 30418964 | ||
| BU664913.1 | - | 3 | 503 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HBD| Gene Symbol: HBD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000519.3 | NP_000510.1 | 3 | 684 | ||
| GenBank mRNA | AY034468.1 | - | 3 | 684 | ||
| BC070282.1 | - | 3 | 540 | 30418964 | ||
| BU664913.1 | - | 3 | 503 | |||