siRNA Details
| Gene Symbol: Cyp2a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133657.1 | NP_598418.1 | 4 | 591 | ||
| GenBank mRNA | BC018356.1 | - | 4 | 591 | 4195640 | |
| BC094017.1 | - | 4 | 579 | 5101743 | ||
| L06463.1 | - | 4 | 577 | |||
| Gene Symbol: Cyp2a22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC151116.1 | - | 4 | 596 | 9088029 | |
| BC158034.1 | - | 4 | 596 | 9007337 | ||

Cyp2a12
Genomic Map