siRNA Details
| Gene Symbol: Cyp2b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009999.4 | NP_034129.1 | 7 | 1111 | ||
| GenBank mRNA | AF128849.1 | - | 7 | 1093 | ||
| AF129405.1 | - | 6 | 907 | |||
| AK028103.1 | - | 7 | 1111 | |||
| BC060973.1 | - | 7 | 1105 | 30301385 | ||
| M21856.1 | - | 7 | 1093 | |||
| X99715.1 | - | 7 | 1089 | |||

Cyp2b10
Genomic Map